ENA371464458 Enamine

1-(1-benzothiophen-3-yl)ethan-1-one

  • MDL number MFCD00051638
  • Cas Number : 1128-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-65
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111010
Availability details for
Quantity
  • Availability
  • Add to Cart