ENA371464463 Enamine

N-[(6-methylpyridin-2-yl)methylidene]hydroxylamine

  • Cas Number : 1265324-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111017
Availability details for
Quantity
  • Availability
  • Add to Cart