ENA371464468 Enamine

4-[4-(4-chlorophenyl)piperazin-1-yl]-4-oxobutanoic acid

  • MDL number MFCD03361824
  • Cas Number : 540521-73-1
  • Molecular Weight : 296.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111022
Availability details for
Quantity
  • Availability
  • Add to Cart