ENA371464470 Enamine

3-{[5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl]carbamoyl}propanoic acid

  • MDL number MFCD00842493
  • Cas Number : 1179399-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111024
Availability details for
Quantity
  • Availability
  • Add to Cart