ENA371464474 Enamine

2-N-tert-butylpyridine-2,3-diamine

  • MDL number MFCD10688728
  • Cas Number : 1022146-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111029
Availability details for
Quantity
  • Availability
  • Add to Cart