ENA371464476 Enamine

1-isocyano-2,4-dimethoxybenzene

  • MDL number MFCD04117585
  • Cas Number : 1984-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111031
Availability details for
Quantity
  • Availability
  • Add to Cart