ENA371464482 Enamine

1-isocyanato-2-(propan-2-yl)benzene

  • MDL number MFCD00013850
  • Cas Number : 56309-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 87
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111037

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P284,P285,P301+P310,P302+P352,P304+P340,P304+P341,P305+P351+P338,P310,P312,P320,P330,P332+P313,P337+P313,P342+P311,P362,P403+P233,P405,P501
hcodes T+,Xi
MSDS https://www.enaminestore.com/catalog/EN300-111037
pictograms GHS06,GHS08
hstatements H301,H315,H319,H330,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart