ENA371464485 Enamine

2-(2,3-dihydro-1H-inden-1-yl)propanedioic acid

  • MDL number MFCD00053471
  • Cas Number : 195071-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111040
Availability details for
Quantity
  • Availability
  • Add to Cart