ENA371464493 Enamine

4-(2-methylprop-2-en-1-yl)-5-(3,4,5-trimethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02669448
  • Cas Number : 537016-35-6
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111048
Availability details for
Quantity
  • Availability
  • Add to Cart