ENA371464494 Enamine

2-{[5-benzyl-4-(prop-2-en-1-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD02671609
  • Cas Number : 56929-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111049
Availability details for
Quantity
  • Availability
  • Add to Cart