ENA371464497 Enamine

4-(2-methylprop-2-en-1-yl)-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00619000
  • Cas Number : 328556-78-1
  • Molecular Weight : 261.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-methyl-2-propenyl)-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol
Inchi Code 1S/C13H15N3OS/c1-10(2)8-16-12(14-15-13(16)18)9-17-11-6-4-3-5-7-11/h3-7H,1,8-9H2,2H3,(H,15,18)
Inchi Key TXNWUQMHKIYXJK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111052
Availability details for
Quantity
  • Availability
  • Add to Cart