ENA371464504 Enamine

5-(3-methyl-2-nitrophenyl)-4-(2-methylprop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01408833
  • Cas Number : 328556-89-4
  • Molecular Weight : 290.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(3-methyl-2-nitrophenyl)-4-(2-methyl-2-propenyl)-4H-1,2,4-triazol-3-yl hydrosulfide
Inchi Code 1S/C13H14N4O2S/c1-8(2)7-16-12(14-15-13(16)20)10-6-4-5-9(3)11(10)17(18)19/h4-6H,1,7H2,2-3H3,(H,15,20)
Inchi Key PUBFRHXTIKNIPW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111059
Availability details for
Quantity
  • Availability
  • Add to Cart