ENA371464512 Enamine

N-[(3,5-dimethyl-1-pentyl-1H-pyrazol-4-yl)methylidene]hydroxylamine

  • Cas Number : 307327-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111067
Availability details for
Quantity
  • Availability
  • Add to Cart