ENA371464513 Enamine

2-[(4-amino-5-benzyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetic acid

  • MDL number MFCD02046319
  • Cas Number : 337487-57-7
  • Molecular Weight : 264.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111068
Availability details for
Quantity
  • Availability
  • Add to Cart