ENA371464515 Enamine

N-[(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methylidene]hydroxylamine

  • MDL number MFCD01164028
  • Cas Number : 23890-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111070

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111070
Availability details for
Quantity
  • Availability
  • Add to Cart