ENA371464517 Enamine

4-benzyl-3-(3,4-dimethoxyphenyl)-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD01242676
  • Cas Number : 299921-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111072

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P337+P313,P363,P501
hcodes Xn,Xi
MSDS https://www.enaminestore.com/catalog/EN300-111072
pictograms GHS07
hstatements H302,H312,H319,H332
Availability details for
Quantity
  • Availability
  • Add to Cart