ENA371464524 Enamine

1-(4-chlorophenyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD00543836
  • Cas Number : 55507-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111080

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111080
Availability details for
Quantity
  • Availability
  • Add to Cart