ENA371464536 Enamine

2-(dimethylamino)-6-methylpyrimidine-4-carboxylic acid

  • MDL number MFCD10032942
  • Cas Number : 1018648-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111092
Availability details for
Quantity
  • Availability
  • Add to Cart