ENA371464540 Enamine

2-(2,6-dimethylphenyl)acetic acid

  • MDL number MFCD06208111
  • Cas Number : 938-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111110
Availability details for
Quantity
  • Availability
  • Add to Cart