ENA371464549 Enamine

5-oxo-4-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09035092
  • Cas Number : 937650-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111119
Availability details for
Quantity
  • Availability
  • Add to Cart