ENA371464551 Enamine

5-(4-chlorophenyl)-4-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD13248840
  • Cas Number : 1146222-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-111121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111121
Availability details for
Quantity
  • Availability
  • Add to Cart