ENA371464554 Enamine

3-(1H-1,3-benzodiazol-1-yl)butanoic acid hydrochloride

  • MDL number MFCD08143956
  • Cas Number : 35321-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111124
Availability details for
Quantity
  • Availability
  • Add to Cart