ENA371464555 Enamine

5H,6H,7H-cyclopenta[d]pyrimidin-4-amine

  • MDL number MFCD00603725
  • Cas Number : 26979-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111125
Availability details for
Quantity
  • Availability
  • Add to Cart