ENA371464556 Enamine

7-tert-butyl-2-methyl-3-phenylpyrazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD07636876
  • Cas Number : 887833-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111126

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111126
Availability details for
Quantity
  • Availability
  • Add to Cart