ENA371464560 Enamine

(2R,3S,4R,5R)-oxane-2,3,4,5-tetrol

  • MDL number MFCD00067708
  • Cas Number : 6748-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111130
Availability details for
Quantity
  • Availability
  • Add to Cart