ENA371464561 Enamine

ethyl 2-{imidazo[1,2-a]pyrimidin-2-yl}acetate

  • MDL number MFCD11656487
  • Cas Number : 61571-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111131

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111131
Availability details for
Quantity
  • Availability
  • Add to Cart