ENA371464567 Enamine

2-amino-1-(4-fluorophenyl)ethan-1-one hydrobromide

  • MDL number MFCD01419795
  • Cas Number : 353506-03-3
  • Molecular Weight : 234.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111137
Availability details for
Quantity
  • Availability
  • Add to Cart