ENA371464574 Enamine

3-N-methylpyridine-2,3-diamine

  • MDL number MFCD00490112
  • Cas Number : 56291-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111144

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-111144
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart