ENA371464575 Enamine

N2-(2,4,6-trimethylphenyl)pyridine-2,3-diamine

  • MDL number MFCD11562418
  • Cas Number : 444325-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111145
Availability details for
Quantity
  • Availability
  • Add to Cart