ENA371464576 Enamine

N2-(naphthalen-1-yl)pyridine-2,3-diamine

  • MDL number MFCD10690552
  • Cas Number : 78750-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111146
Availability details for
Quantity
  • Availability
  • Add to Cart