ENA371464579 Enamine

N2-(4-fluorophenyl)pyridine-2,3-diamine

  • MDL number MFCD11201955
  • Cas Number : 41010-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111149
Availability details for
Quantity
  • Availability
  • Add to Cart