ENA371464588 Enamine

N-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-2-chloroacetamide

  • MDL number MFCD00500657
  • Cas Number : 28242-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110723
Availability details for
Quantity
  • Availability
  • Add to Cart