ENA371464593 Enamine

1-[10-(pyridin-1-ium-1-yl)decyl]pyridin-1-ium dibromide

  • MDL number MFCD00031922
  • Cas Number : 6266-40-6
  • Molecular Weight : 458.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[10-(1-pyridiniumyl)decyl]pyridinium dibromide
Inchi Code 1S/C20H30N2.2BrH/c1(3-5-9-15-21-17-11-7-12-18-21)2-4-6-10-16-22-19-13-8-14-20-22;;/h7-8,11-14,17-20H,1-6,9-10,15-16H2;2*1H/q+2;;/p-2
Inchi Key XZZPVHYTBRRMNM-UHFFFAOYSA-L

Description

Purity 95
Salt Data [Br-]
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111175
Availability details for
Quantity
  • Availability
  • Add to Cart