ENA371464601 Enamine

5,7-difluoro-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD08544288
  • Cas Number : 939758-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111217
Availability details for
Quantity
  • Availability
  • Add to Cart