ENA371464611 Enamine

4-phenylbutanoyl chloride

  • MDL number MFCD00798080
  • Cas Number : 18496-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110725

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-110725
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart