ENA371464614 Enamine

phenyl N-(4-hydroxyphenyl)-N-methylcarbamate

  • MDL number MFCD00586121
  • Cas Number : 955948-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111288
Availability details for
Quantity
  • Availability
  • Add to Cart