ENA371464619 Enamine

4-(4-chlorophenyl)piperazin-1-amine

  • MDL number MFCD02271426
  • Cas Number : 71416-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110727
Availability details for
Quantity
  • Availability
  • Add to Cart