ENA371464625 Enamine

2-(4-nitrophenyl)oxirane

  • MDL number MFCD00181340
  • Cas Number : 6388-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-16699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16699
Availability details for
Quantity
  • Availability
  • Add to Cart