ENA371464634 Enamine

N-(4-hydroxyphenyl)-N,2,2-trimethylpropanamide

  • MDL number MFCD11167639
  • Cas Number : 1038215-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22232
Availability details for
Quantity
  • Availability
  • Add to Cart