ENA371464635 Enamine

4-(4-bromophenyl)-3-(pyridin-4-yl)-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD02938939
  • Cas Number : 117080-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23554
Availability details for
Quantity
  • Availability
  • Add to Cart