ENA371464636 Enamine

1-{5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-yl}ethan-1-amine

  • MDL number MFCD22378799
  • Cas Number : 1394042-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27381
Availability details for
Quantity
  • Availability
  • Add to Cart