ENA371464637 Enamine

1-(9H-fluoren-9-yl)piperazine

  • MDL number MFCD00230324
  • Cas Number : 129604-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27691
Availability details for
Quantity
  • Availability
  • Add to Cart