ENA371464658 Enamine

3-(diethoxymethyl)-2-ethoxyoxolane

  • MDL number MFCD08558978
  • Cas Number : 177940-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51358
Availability details for
Quantity
  • Availability
  • Add to Cart