ENA371464663 Enamine

4-chloro-6-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD10697160
  • Cas Number : 30129-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54584

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54584
Availability details for
Quantity
  • Availability
  • Add to Cart