ENA371464666 Enamine

4-[(tert-butoxy)carbonyl]morpholine-2-carboxylic acid

  • MDL number MFCD01321006
  • Cas Number : 189321-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56371

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-56371
pictograms GHS07,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart