ENA371464677 Enamine

1-aminopentan-3-ol

  • MDL number MFCD12190106
  • Cas Number : 291535-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62209

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-62209
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart