ENA371464684 Enamine

2-chloro-5-(trifluoromethyl)benzene-1-sulfonamide

  • MDL number MFCD00099222
  • Cas Number : 779-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68771
Availability details for
Quantity
  • Availability
  • Add to Cart