ENA371464694 Enamine

thiolane-3-carboxylic acid

  • MDL number MFCD11847784
  • Cas Number : 18133-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-62
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76346
Availability details for
Quantity
  • Availability
  • Add to Cart