ENA371464712 Enamine

3-(chloromethyl)-1-cyclopentyl-1H-pyrazole

  • MDL number MFCD18262439
  • Cas Number : 1260659-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84084
Availability details for
Quantity
  • Availability
  • Add to Cart