ENA371464714 Enamine

2-(6-fluoropyridin-2-yl)acetonitrile

  • MDL number MFCD09924020
  • Cas Number : 312325-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84454
Availability details for
Quantity
  • Availability
  • Add to Cart